Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683229.3 | XP_005683286.1 | 5 - 6 | 759 | 98 | |
| XM_018050107.1 | XP_017905596.1 | 4 - 5 | 535 | 98 | ||
| XM_018050108.1 | XP_017905597.1 | 4 - 5 | 334 | 98 | ||
More Information
Set Membership: |
|

THG1L
Genomic Map