Assay Details
| Gene Symbol: ADAMDEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052342.1 | XP_017907831.1 | 12 - 13 | 1396 | 64 | |
| GenBank mRNA | JO609505.2 | - | 2 - 3 | 474 | 64 | |
More Information
Set Membership: |
|

ADAMDEC1
Genomic Map