Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684796.3 | XP_005684853.1 | 5 - 6 | 1490 | 64 | |
| XM_005684797.3 | XP_005684854.1 | 5 - 6 | 1269 | 64 | ||
| GenBank mRNA | JO594172.2 | - | 5 - 6 | 641 | 64 | |
More Information
Set Membership: |
|

MTFR2
Genomic Map