Assay Details
| Gene Symbol: DPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686036.3 | XP_005686093.1 | 6 - 7 | 629 | 62 | |
| GenBank mRNA | JO616391.2 | - | 2 - 3 | 135 | 62 | |
More Information
Set Membership: |
|

DPF3
Genomic Map