Assay Details
| Gene Symbol: ECHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057003.1 | XP_017912492.1 | 3 - 4 | 613 | 102 | |
| XM_018057004.1 | XP_017912493.1 | 3 - 4 | 617 | 102 | ||
| XM_018057005.1 | XP_017912494.1 | 3 - 4 | 616 | 102 | ||
| GenBank mRNA | JO425181.1 | - | 3 - 4 | 653 | 102 | |
More Information
Set Membership: |
|

ECHDC3
Genomic Map