Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057330.1 | XP_017912819.1 | 3 - 4 | 1054 | 90 | |
| XM_018057331.1 | XP_017912820.1 | 4 - 5 | 1122 | 90 | ||
| GenBank mRNA | KA318917.1 | - | 1 - 2 | 1871 | 90 | |
| KU041749.1 | - | 3 - 4 | 807 | 90 | ||
More Information
Set Membership: |
|

KLF6
Genomic Map