Assay Details
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688653.3 | XP_005688710.2 | 1 - 2 | 194 | 74 | |
GenBank mRNA | JO441433.2 | - | 8 - 9 | 848 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
