Assay Details
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689251.3 | XP_005689308.1 | 8 - 9 | 790 | 67 | |
GenBank mRNA | JO427729.1 | - | 8 - 9 | 856 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
