Assay Details
| Gene Symbol: ELP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059295.1 | XP_017914784.1 | 8 - 9 | 1075 | 73 | |
| XM_018059296.1 | XP_017914785.1 | 8 - 9 | 1074 | 73 | ||
More Information
Set Membership: |
|

ELP4
Genomic Map