Assay Details
| Gene Symbol: LOC102190340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059200.1 | XP_017914689.1 | 4 - 5 | 761 | 79 | |
| XM_018059201.1 | XP_017914690.1 | 4 - 5 | 533 | 79 | ||
| XM_018059202.1 | XP_017914691.1 | 4 - 5 | 761 | 79 | ||
| Gene Symbol: LOC108637651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JT698282.1 | - | 3 - 4 | 494 | 79 | |
More Information
Set Membership: |
|

LOC102190340
Genomic Map