Assay Details
| Gene Symbol: NEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691024.3 | XP_005691081.1 | 6 - 7 | 1089 | 71 | |
| GenBank mRNA | JO613507.2 | - | 2 - 3 | 850 | 71 | |
More Information
Set Membership: |
|

NEK2
Genomic Map