Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694015.2 | XP_005694072.1 | 6 - 7 | 817 | 66 | |
| XM_005694016.2 | XP_005694073.1 | 6 - 7 | 816 | 66 | ||
| GenBank mRNA | JO324308.2 | - | 6 - 7 | 775 | 66 | |
| JO610405.2 | - | 6 - 7 | 807 | 66 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map