Assay Details
| Gene Symbol: C23H6orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696592.3 | XP_005696649.2 | 1 | 1631 | 75 | |
| GenBank mRNA | JT684993.1 | - | 1 | 1466 | 75 | |
More Information
Set Membership: |
|

C23H6orf47
Genomic Map