Assay Details
| Gene Symbol: LOC102170569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696788.2 | XP_005696845.1 | 3 - 4 | 401 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102170569| Gene Symbol: LOC102170569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696788.2 | XP_005696845.1 | 3 - 4 | 401 | 80 | |
Set Membership: |
|