Assay Details
| Gene Symbol: PSMA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039666.1 | XP_017895155.1 | 4 - 5 | 427 | 66 | |
| XM_018039668.1 | XP_017895157.1 | 4 - 5 | 427 | 66 | ||
| XM_018039669.1 | XP_017895158.1 | 4 - 5 | 427 | 66 | ||
More Information
Set Membership: |
|

PSMA8
Genomic Map