Assay Details
| Gene Symbol: LOC100861263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI861626.1 | - | 9 - 10 | 768 | 75 | |
| JO633195.2 | - | 9 - 10 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100861263| Gene Symbol: LOC100861263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI861626.1 | - | 9 - 10 | 768 | 75 | |
| JO633195.2 | - | 9 - 10 | 75 | |||
Set Membership: |
|