Assay Details
| Gene Symbol: LOC100861263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697593.3 | XP_005697650.1 | 6 - 7 | 638 | 80 | |
| GenBank mRNA | JI861626.1 | - | 6 - 7 | 516 | 80 | |
| JO633195.2 | - | 6 - 7 | 697 | 80 | ||
More Information
Set Membership: |
|

LOC100861263
Genomic Map