Assay Details
| Gene Symbol: LOC100861263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697593.3 | XP_005697650.1 | 8 - 9 | 795 | 88 | |
| GenBank mRNA | JO633195.2 | - | 8 - 9 | 854 | 88 | |
More Information
Set Membership: |
|

LOC100861263
Genomic Map