Assay Details
| Gene Symbol: LOC102183750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697657.3 | XP_005697714.1 | 8 - 9 | 1474 | 69 | |
| XM_018040158.1 | XP_017895647.1 | 8 - 9 | 1513 | 69 | ||
| XM_018040159.1 | XP_017895648.1 | 8 - 9 | 1473 | 69 | ||
| XM_018040161.1 | XP_017895650.1 | 7 - 8 | 1266 | 69 | ||
| XM_018040162.1 | XP_017895651.1 | 7 - 8 | 1303 | 69 | ||
| XM_018040163.1 | XP_017895652.1 | 7 - 8 | 1263 | 69 | ||
| GenBank mRNA | JO582012.1 | - | 8 - 9 | 1514 | 69 | |
| JO582013.1 | - | 8 - 9 | 1550 | 69 | ||
More Information
Set Membership: |
|

LOC102183750
Genomic Map