Assay Details
| Gene Symbol: LOC108635301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045508.1 | XP_017900997.1 | 2 - 3 | 407 | 89 | |
| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048932.1 | XP_017904421.1 | 1 - 2 | 308 | 89 | |
| GenBank mRNA | JO337979.2 | - | 5 - 6 | 721 | 89 | |
| JO640929.2 | - | 1 - 2 | 420 | 89 | ||
More Information
Set Membership: |
|

LOC108635301
Genomic Map