Assay Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049980.1 | XP_017905469.1 | 10 - 11 | 1083 | 70 | |
| XM_018049981.1 | XP_017905470.1 | 9 - 10 | 798 | 70 | ||
More Information
Set Membership: |
|

CAMK4
Genomic Map