Assay Details
Gene Symbol: ZNF462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052556.1 | XP_017908045.1 | 6 - 7 | 6544 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
