Assay Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053181.1 | XP_017908670.1 | 2 - 3 | 393 | 74 | |
| XM_018053182.1 | XP_017908671.1 | 2 - 3 | 323 | 74 | ||
More Information
Set Membership: |
|

MRAP2
Genomic Map