Assay Details
| Gene Symbol: PROC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065804.1 | XP_017921293.1 | 9 - 10 | 1050 | 63 | |
| XM_018065808.1 | XP_017921297.1 | 8 - 9 | 912 | 63 | ||
| XM_018065812.1 | XP_017921301.1 | 8 - 9 | 899 | 63 | ||
| XM_018065813.1 | XP_017921302.1 | 10 - 11 | 1095 | 63 | ||
| XM_018065814.1 | XP_017921303.1 | 9 - 10 | 964 | 63 | ||
| XM_018065815.1 | XP_017921304.1 | 10 - 11 | 1091 | 63 | ||
| XM_018065818.1 | XP_017921307.1 | 9 - 10 | 960 | 63 | ||
| XM_018065820.1 | XP_017921309.1 | 8 - 9 | 986 | 63 | ||
| GenBank mRNA | JO421087.2 | - | 7 - 8 | 1302 | 63 | |
More Information
Set Membership: |
|

PROC
Genomic Map