Assay Details
| Gene Symbol: LOC102182418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056311.1 | XP_017911800.1 | 10 - 11 | 1794 | 76 | |
| GenBank mRNA | JO407966.2 | - | 4 - 5 | 516 | 76 | |
More Information
Set Membership: |
|

LOC102182418
Genomic Map