Assay Details
| Gene Symbol: LOC102182418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056311.1 | XP_017911800.1 | 8 - 9 | 1493 | 68 | |
| GenBank mRNA | JO407966.2 | - | 2 - 3 | 197 | 68 | |
More Information
Set Membership: |
|

LOC102182418
Genomic Map