Assay Details
| Gene Symbol: SCN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063994.1 | XP_017919483.1 | 11 - 12 | 1953 | 66 | |
| XM_018063997.1 | XP_017919486.1 | 11 - 12 | 2008 | 66 | ||
| XM_018064002.1 | XP_017919491.1 | 11 - 12 | 1871 | 66 | ||
| XM_018064007.1 | XP_017919496.1 | 11 - 12 | 1944 | 66 | ||
More Information
Set Membership: |
|

SCN2A
Genomic Map