Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060148.1 | XP_017915637.1 | 3 - 4 | 1272 | 91 | |
| XM_018060149.1 | XP_017915638.1 | 3 - 4 | 990 | 91 | ||
| XM_018060150.1 | XP_017915639.1 | 3 - 4 | 1272 | 91 | ||
| GenBank mRNA | JO603360.2 | - | 3 - 4 | 973 | 91 | |
More Information
Set Membership: |
|

RO60
Genomic Map