Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060148.1 | XP_017915637.1 | 8 - 9 | 1934 | 83 | |
| XM_018060149.1 | XP_017915638.1 | 8 - 9 | 1652 | 83 | ||
| XM_018060150.1 | XP_017915639.1 | 7 - 8 | 1817 | 83 | ||
| GenBank mRNA | JO603360.2 | - | 8 - 9 | 1635 | 83 | |
| KA320210.1 | - | 1 - 2 | 51 | 83 | ||
More Information
Set Membership: |
|

RO60
Genomic Map