Assay Details
| Gene Symbol: C19H17orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971785.1 | XP_013827239.1 | 2 - 3 | 1664 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19H17orf47| Gene Symbol: C19H17orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971785.1 | XP_013827239.1 | 2 - 3 | 1664 | 84 | |
Set Membership: |
|