Assay Details
| Gene Symbol: NARF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065398.1 | XP_017920887.1 | 3 - 4 | 379 | 73 | |
| XM_018065399.1 | XP_017920888.1 | 3 - 4 | 382 | 73 | ||
More Information
Set Membership: |
|

NARF
Genomic Map