Assay Details
| Gene Symbol: SLCO3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066138.1 | XP_017921627.1 | 4 - 5 | 1105 | 61 | |
| XM_018066139.1 | XP_017921628.1 | 4 - 5 | 1105 | 61 | ||
| GenBank mRNA | JO637541.2 | - | 4 - 5 | 1047 | 61 | |
More Information
Set Membership: |
|

SLCO3A1
Genomic Map