Assay Details
| Gene Symbol: LOC106503400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044458.1 | XP_017899947.1 | 3 - 4 | 549 | 73 | |
| GenBank mRNA | JO538910.2 | - | 5 - 6 | 891 | 73 | |
More Information
Set Membership: |
|

LOC106503400
Genomic Map