Assay Details
| Gene Symbol: TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038924.1 | XP_017894413.1 | 9 - 10 | 1249 | 57 | |
| GenBank mRNA | JO413496.2 | - | 1 - 2 | 1428 | 57 | |
| JO424394.1 | - | 9 - 10 | 1187 | 57 | ||
More Information
Set Membership: |
|

TXNDC5
Genomic Map