Assay Details
| Gene Symbol: SEPT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040097.1 | XP_017895586.1 | 3 - 4 | 377 | 77 | |
| XM_018040098.1 | XP_017895587.1 | 2 - 3 | 284 | 77 | ||
More Information
Set Membership: |
|

SEPT12
Genomic Map