Assay Details
| Gene Symbol: SPINDOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043364.1 | XP_017898853.1 | 3 - 4 | 2728 | 80 | |
| XM_018043365.1 | XP_017898854.1 | 3 - 4 | 2730 | 80 | ||
| XM_018043366.1 | XP_017898855.1 | 4 - 5 | 1001 | 80 | ||
| GenBank mRNA | JO418481.2 | - | 4 - 5 | 676 | 80 | |
| JO646621.2 | - | 4 - 5 | 929 | 80 | ||
More Information
Set Membership: |
|

SPINDOC
Genomic Map