Assay Details
Gene Symbol: SYT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043560.1 | XP_017899049.1 | 5 - 6 | 1184 | 58 | |
XM_018043561.1 | XP_017899050.1 | 6 - 7 | 1583 | 58 | ||
GenBank mRNA | JO639681.2 | - | 6 - 7 | 1113 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
