Assay Details
| Gene Symbol: LOC102188566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042764.1 | XP_017898253.1 | 1 - 2 | 899 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102188566| Gene Symbol: LOC102188566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042764.1 | XP_017898253.1 | 1 - 2 | 899 | 121 | |
Set Membership: |
|