Assay Details
| Gene Symbol: LOC102188566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042764.1 | XP_017898253.1 | 4 - 5 | 1336 | 100 | |
| GenBank mRNA | KA344819.1 | - | 1 - 2 | 180 | 100 | |
More Information
Set Membership: |
|

LOC102188566
Genomic Map