Assay Details
| Gene Symbol: LOC102187632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676339.3 | XP_005676396.1 | 21 - 22 | 2685 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102187632| Gene Symbol: LOC102187632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676339.3 | XP_005676396.1 | 21 - 22 | 2685 | 98 | |
Set Membership: |
|