Assay Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046236.1 | XP_017901725.1 | 12 - 13 | 1636 | 64 | |
| XM_018046237.1 | XP_017901726.1 | 12 - 13 | 1633 | 64 | ||
| XM_018046238.1 | XP_017901727.1 | 12 - 13 | 1516 | 64 | ||
| XM_018046239.1 | XP_017901728.1 | 12 - 13 | 1513 | 64 | ||
| XM_018046240.1 | XP_017901729.1 | 11 - 12 | 1554 | 64 | ||
More Information
Set Membership: |
|

CLK2
Genomic Map