Assay Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680565.3 | XP_005680622.1 | 2 - 3 | 728 | 74 | |
| XM_018048196.1 | XP_017903685.1 | 2 - 3 | 713 | 74 | ||
| GenBank mRNA | JO624641.2 | - | 2 - 3 | 526 | 74 | |
More Information
Set Membership: |
|

CHST11
Genomic Map