Assay Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683509.3 | XP_005683566.1 | 1 - 2 | 386 | 80 | |
| XM_013965831.2 | XP_013821285.1 | 1 - 2 | 386 | 80 | ||
More Information
Set Membership: |
|

AADAT
Genomic Map