Assay Details
| Gene Symbol: AP5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685905.3 | XP_005685962.2 | 3 - 4 | 984 | 70 | |
| XM_005685906.3 | XP_005685963.2 | 3 - 4 | 1193 | 70 | ||
More Information
Set Membership: |
|

AP5M1
Genomic Map