Assay Details
| Gene Symbol: EDNRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687682.3 | XP_005687739.1 | 6 - 7 | 1459 | 65 | |
| GenBank mRNA | JT700244.1 | - | 4 - 5 | 802 | 65 | |
More Information
Set Membership: |
|

EDNRB
Genomic Map