Assay Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688773.3 | XP_005688830.3 | 2 - 3 | 563 | 86 | |
| XM_018058789.1 | XP_017914278.1 | 3 - 4 | 218 | 86 | ||
| XM_018058790.1 | XP_017914279.1 | 3 - 4 | 218 | 86 | ||
| XM_018058791.1 | XP_017914280.1 | 2 - 3 | 155 | 86 | ||
| XM_018058792.1 | XP_017914281.1 | 3 - 4 | 502 | 86 | ||
| XM_018058793.1 | XP_017914282.1 | 2 - 3 | 449 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018058791.1, XM_018058792.1, XM_018058793.1 transcripts.
Set Membership: |
|

ZNF7
Genomic Map