Assay Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688773.3 | XP_005688830.3 | 3 - 4 | 689 | 62 | |
| XM_018058789.1 | XP_017914278.1 | 4 - 5 | 344 | 62 | ||
| XM_018058791.1 | XP_017914280.1 | 3 - 4 | 281 | 62 | ||
| XM_018058792.1 | XP_017914281.1 | 4 - 5 | 628 | 62 | ||
| XM_018058793.1 | XP_017914282.1 | 3 - 4 | 575 | 62 | ||
| GenBank mRNA | JT682753.1 | - | 3 - 4 | 283 | 62 | |
More Information
Set Membership: |
|

ZNF7
Genomic Map