Assay Details
| Gene Symbol: NLRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689731.3 | XP_005689788.2 | 8 - 9 | 2865 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP14| Gene Symbol: NLRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689731.3 | XP_005689788.2 | 8 - 9 | 2865 | 65 | |
Set Membership: |
|