Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690037.3 | XP_005690094.1 | 4 - 5 | 394 | 80 | |
| XM_005690038.3 | XP_005690095.1 | 5 - 6 | 654 | 80 | ||
| XM_005690039.3 | XP_005690096.1 | 4 - 5 | 386 | 80 | ||
| XM_005690040.3 | XP_005690097.1 | 3 - 4 | 336 | 80 | ||
| XM_005690041.2 | XP_005690098.1 | 4 - 5 | 332 | 80 | ||
| XM_005690043.3 | XP_005690100.1 | 4 - 5 | 596 | 80 | ||
| XM_013969799.2 | XP_013825253.1 | 4 - 5 | 324 | 80 | ||
| GenBank mRNA | JO374272.2 | - | 5 - 6 | 441 | 80 | |
| JO446293.2 | - | 4 - 5 | 453 | 80 | ||
More Information
Set Membership: |
|

IMMP1L
Genomic Map