Assay Details
| Gene Symbol: LOC102187615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060360.1 | XP_017915849.1 | 4 - 5 | 387 | 94 | |
| GenBank mRNA | JO585116.2 | - | 4 - 5 | 355 | 94 | |
| JO647116.2 | - | 1 - 2 | 429 | 94 | ||
More Information
Set Membership: |
|

LOC102187615
Genomic Map